siRNA Details
| Gene Symbol: Gm19744 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK032387.1 | - | 4 | 3662 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CO427011.1 | - | 2 | 458 | Chr. 8 - 48864186 - 48869310 [+] on Build GRCm38 | |

Gm19744
Genomic Map