siRNA Details
| Gene Symbol: 0610009F21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK002391.1 | - | 1 | 24 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY702926.1 | - | 1 | 22 | Chr. 16 - 91947327 - 91947788 [+] on Build GRCm38 | |

0610009F21Rik
Genomic Map