siRNA Details
| Gene Symbol: 4930556M19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045065.1 | 4 | 700 | |||
| GenBank mRNA | AK047371.1 | - | 2 | 2467 | ||
| AK047938.1 | - | 2 | 1563 | |||
| BY740872.1 | - | 4 | 700 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CB244482.1 | - | 2 | 255 | Chr. 15 - 10719358 - 10721350 [+] on Build GRCm38 | |

4930556M19Rik
Genomic Map