siRNA Details
| Gene Symbol: 4930556M19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045065.1 | 4 | 1650 | |||
| GenBank mRNA | AA178715.1 | - | 1 | 125 | ||
| AI592271.1 | - | 1 | 126 | |||
| AK047371.1 | - | 2 | 3418 | |||
| AK047938.1 | - | 2 | 2513 | |||
| BB344878.2 | - | 1 | 71 | |||
| BB364664.2 | - | 1 | 624 | |||

4930556M19Rik
Genomic Map