siRNA Details
| Gene Symbol: 4930556M19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045065.1 | 4 | 1345 | |||
| GenBank mRNA | AK047371.1 | - | 2 | 3113 | ||
| AK047938.1 | - | 2 | 2208 | |||
| BB364664.2 | - | 1 | 319 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930556M19Rik| Gene Symbol: 4930556M19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045065.1 | 4 | 1345 | |||
| GenBank mRNA | AK047371.1 | - | 2 | 3113 | ||
| AK047938.1 | - | 2 | 2208 | |||
| BB364664.2 | - | 1 | 319 | |||