siRNA Details
| Gene Symbol: 4930556M19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045065.1 | 2, 3 | 163 | |||
| GenBank mRNA | AK047371.1 | - | 1 | 1930 | ||
| AK047938.1 | - | 1 | 1026 | |||
| BY740872.1 | - | 2, 3 | 163 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930556M19Rik| Gene Symbol: 4930556M19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045065.1 | 2, 3 | 163 | |||
| GenBank mRNA | AK047371.1 | - | 1 | 1930 | ||
| AK047938.1 | - | 1 | 1026 | |||
| BY740872.1 | - | 2, 3 | 163 | |||