siRNA Details
| Gene Symbol: 4930556M19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK047371.1 | - | 1 | 939 | ||
| AK047938.1 | - | 1 | 35 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930556M19Rik| Gene Symbol: 4930556M19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK047371.1 | - | 1 | 939 | ||
| AK047938.1 | - | 1 | 35 | |||