siRNA Details
| Gene Symbol: Gm38414 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK039009.1 | - | 2, 3 | 195 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB632808.1 | - | 2, 3 | 195 | Chr. 8 - 36159479 - 36160804 [+] on Build GRCm38 | |

Gm38414
Genomic Map