siRNA Details
| Gene Symbol: C230079O03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040459.1 | 5 | 1244 | |||
| GenBank mRNA | AK048899.1 | - | 5 | 1245 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C230079O03Rik| Gene Symbol: C230079O03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040459.1 | 5 | 1244 | |||
| GenBank mRNA | AK048899.1 | - | 5 | 1245 | ||