siRNA Details
| Gene Symbol: C230079O03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040459.1 | 4 | 320 | |||
| GenBank mRNA | AK048899.1 | - | 4 | 321 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB651410.1 | - | 4 | 321 | Chr. 7 - 136332993 - 136349415 [-] on Build GRCm38 | |

C230079O03Rik
Genomic Map