siRNA Details
| Gene Symbol: Hells | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021390.1 | - | 277 | |||
| AK043271.1 | - | 502 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hells| Gene Symbol: Hells | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021390.1 | - | 277 | |||
| AK043271.1 | - | 502 | ||||