siRNA Details
| Gene Symbol: E130118H10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021395.1 | - | 1 | 297 | ||
| AK165039.1 | - | 1 | 5461 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY731372.1 | - | 1 | 295 | Chr. 5 - 31572389 - 31573011 [+] on Build GRCm38 | |

E130118H10Rik
Genomic Map