siRNA Details
| Gene Symbol: E130119H09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021396.1 | - | 3 | 512 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY731386.1 | - | 3 | 512 | Chr. 13 - 53912644 - 53915748 [-] on Build GRCm38 | |

E130119H09Rik
Genomic Map