siRNA Details
| Gene Symbol: Peli3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK135575.1 | - | 6 | 2738 | ||
| BC022735.1 | - | 1 | 159 | |||
| BC057046.1 | - | 1 | 3112 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG297260.1 | - | 158 | Chr. - - [-] on Build | ||

Peli3
Genomic Map