siRNA Details
Gene Symbol: Peli3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020060.1 | - | 1 | 814 | ||
BC022735.1 | - | 1 | 1296 | |||
BC057046.1 | - | 1 | 4249 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Peli3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020060.1 | - | 1 | 814 | ||
BC022735.1 | - | 1 | 1296 | |||
BC057046.1 | - | 1 | 4249 |