siRNA Details
| Gene Symbol: Peli3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020060.1 | - | 1 | 763 | ||
| BC022735.1 | - | 1 | 1245 | |||
| BC057046.1 | - | 1 | 4198 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Peli3| Gene Symbol: Peli3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020060.1 | - | 1 | 763 | ||
| BC022735.1 | - | 1 | 1245 | |||
| BC057046.1 | - | 1 | 4198 | |||