siRNA Details
| Gene Symbol: Peli3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020060.1 | - | 1 | 100 | ||
| BC022735.1 | - | 1 | 581 | |||
| BC057046.1 | - | 1 | 3534 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG297260.1 | - | 581 | Chr. - - [-] on Build | ||

Peli3
Genomic Map