siRNA Details
| Gene Symbol: Klrb1-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073569.1 | 2, 3 | 192 | |||
| GenBank mRNA | DQ237930.1 | - | 2, 3 | 192 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klrb1-ps1| Gene Symbol: Klrb1-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_073569.1 | 2, 3 | 192 | |||
| GenBank mRNA | DQ237930.1 | - | 2, 3 | 192 | ||