siRNA Details
| Gene Symbol: Gm15328 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045399.1 | 3 | 2865 | |||
| GenBank mRNA | AK085921.1 | - | 3 | 2866 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm15328| Gene Symbol: Gm15328 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045399.1 | 3 | 2865 | |||
| GenBank mRNA | AK085921.1 | - | 3 | 2866 | ||