siRNA Details
| Gene Symbol: Gm15328 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045399.1 | 3 | 414 | |||
| GenBank mRNA | AK085921.1 | - | 3 | 415 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB662184.1 | - | 3 | 410 | Chr. 18 - 16816411 - 16820444 [+] on Build GRCm38 | |

Gm15328
Genomic Map