siRNA Details
| Gene Symbol: Zmynd19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK086718.1 | - | 755 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CD807049.1 | - | 1 | 22 | Chr. 2 - 24960198 - 24960931 [+] on Build GRCm38 | |

Zmynd19
Genomic Map