siRNA Details
| Gene Symbol: Gm34885 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133341.1 | - | 3 | 514 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CJ047701.1 | - | 3 | 514 | Chr. 9 - 37312136 - 37321039 [+] on Build GRCm38 | |

Gm34885
Genomic Map