siRNA Details
| Gene Symbol: Bcorl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020114.1 | - | 3 | 1096 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcorl1| Gene Symbol: Bcorl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020114.1 | - | 3 | 1096 | ||