siRNA Details
| Gene Symbol: 2610020C07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011479.1 | - | 2 | 688 | ||
| AK029005.1 | - | 2 | 697 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2610020C07Rik| Gene Symbol: 2610020C07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011479.1 | - | 2 | 688 | ||
| AK029005.1 | - | 2 | 697 | |||