siRNA Details
| Gene Symbol: 4930405D11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045956.1 | 1 | 43 | |||
| GenBank mRNA | AK147079.1 | - | 1 | 44 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY740750.1 | - | 1 | 42 | Chr. 11 - 90804692 - 90895149 [+] on Build GRCm38 | |

4930405D11Rik
Genomic Map