siRNA Details
| Gene Symbol: 4930405D11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045956.1 | 2 | 134 | |||
| GenBank mRNA | AK147079.1 | - | 2 | 135 | ||
| Gene Symbol: 4731417B20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK014627.1 | - | 1 | 25 | ||

4930405D11Rik
Genomic Map