siRNA Details
| Gene Symbol: 1700007F19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040538.1 | 6 | 669 | |||
| GenBank mRNA | AK005707.1 | - | 6 | 650 | ||
| AK017083.1 | - | 6 | 671 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700007F19Rik| Gene Symbol: 1700007F19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040538.1 | 6 | 669 | |||
| GenBank mRNA | AK005707.1 | - | 6 | 650 | ||
| AK017083.1 | - | 6 | 671 | |||