siRNA Details
| Gene Symbol: Ybey | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK051909.1 | - | 1 | 397 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB657821.1 | - | 1 | 394 | Chr. 10 - 76468239 - 76468861 [-] on Build GRCm38 | |

Ybey
Genomic Map