siRNA Details
| Gene Symbol: Tomm6os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045945.1 | 3 | 2318 | |||
| GenBank mRNA | AK046990.1 | - | 3 | 2320 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tomm6os| Gene Symbol: Tomm6os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045945.1 | 3 | 2318 | |||
| GenBank mRNA | AK046990.1 | - | 3 | 2320 | ||