siRNA Details
| Gene Symbol: 4933428L12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016971.1 | - | 1 | 1298 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | EL962590.1 | - | 1 | 515 | Chr. 14 - 24447340 - 24447904 [+] on Build GRCm38 | |

4933428L12Rik
Genomic Map