siRNA Details
Gene Symbol: 4933428L01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016970.1 | - | 1 | 372 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716653.1 | - | 1 | 368 | Chr. 15 - 43700336 - 43701032 [-] on Build GRCm38 |
