siRNA Details
| Gene Symbol: Gm19345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043814.1 | - | 4 | 617 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BM935040.1 | - | 4 | 448 | Chr. 7 - 19852172 - 19858109 [+] on Build GRCm38 | |

Gm19345
Genomic Map