siRNA Details
| Gene Symbol: Gm6602 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045362.1 | 1 | 274 | |||
| GenBank mRNA | AK090313.1 | - | 1 | 275 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm6602| Gene Symbol: Gm6602 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045362.1 | 1 | 274 | |||
| GenBank mRNA | AK090313.1 | - | 1 | 275 | ||