siRNA Details
Gene Symbol: Gm1720 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046015.1 | 9 | 1261 | |||
GenBank mRNA | AK144520.1 | - | 9 | 1262 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm1720 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046015.1 | 9 | 1261 | |||
GenBank mRNA | AK144520.1 | - | 9 | 1262 |