siRNA Details
| Gene Symbol: 4930513D17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045416.1 | 2, 3 | 193 | |||
| GenBank mRNA | AK015779.1 | - | 2, 3 | 193 | ||
| AK132902.1 | - | 2, 3 | 152 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930513D17Rik| Gene Symbol: 4930513D17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045416.1 | 2, 3 | 193 | |||
| GenBank mRNA | AK015779.1 | - | 2, 3 | 193 | ||
| AK132902.1 | - | 2, 3 | 152 | |||