siRNA Details
| Gene Symbol: 3110033O22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019423.1 | - | 1 | 39 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

3110033O22Rik| Gene Symbol: 3110033O22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019423.1 | - | 1 | 39 | ||