siRNA Details
| Gene Symbol: 4930532M18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108050.1 | 3 | 197 | |||
| GenBank mRNA | AK015950.1 | - | 3 | 199 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930532M18Rik| Gene Symbol: 4930532M18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108050.1 | 3 | 197 | |||
| GenBank mRNA | AK015950.1 | - | 3 | 199 | ||