siRNA Details
| Gene Symbol: 4930532M18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108050.1 | 1 | 58 | |||
| GenBank mRNA | AK015950.1 | - | 1 | 60 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715514.1 | - | 1 | 59 | Chr. 1 - 154255689 - 154270418 [+] on Build GRCm38 | |

4930532M18Rik
Genomic Map