siRNA Details
| Gene Symbol: 4930533B01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040614.1 | 4 | 585 | |||
| GenBank mRNA | AK015951.1 | - | 4 | 586 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715515.1 | - | 4 | 583 | Chr. 2 - 113691148 - 113708548 [-] on Build GRCm38 | |

4930533B01Rik
Genomic Map