siRNA Details
| Gene Symbol: 4930533B01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040614.1 | 6 | 861 | |||
| GenBank mRNA | AK015951.1 | - | 6 | 862 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930533B01Rik| Gene Symbol: 4930533B01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040614.1 | 6 | 861 | |||
| GenBank mRNA | AK015951.1 | - | 6 | 862 | ||