siRNA Details
Gene Symbol: 4930533B01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040614.1 | 6 | 915 | |||
GenBank mRNA | AK015951.1 | - | 6 | 916 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930533B01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040614.1 | 6 | 915 | |||
GenBank mRNA | AK015951.1 | - | 6 | 916 |