siRNA Details
| Gene Symbol: 4930578E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045391.1 | 6 | 1414 | |||
| GenBank mRNA | AK132929.1 | - | 6 | 1415 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930578E11Rik| Gene Symbol: 4930578E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045391.1 | 6 | 1414 | |||
| GenBank mRNA | AK132929.1 | - | 6 | 1415 | ||