siRNA Details
| Gene Symbol: 4930578E11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045391.1 | 3 | 1006 | |||
| GenBank mRNA | AK019814.1 | - | 2 | 376 | ||
| AK132929.1 | - | 3 | 1007 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU605115.1 | - | 3 | 422 | Chr. 18 - 29451745 - 29495832 [-] on Build GRCm38 | |

4930578E11Rik
Genomic Map