siRNA Details
| Gene Symbol: Dtna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008850.1 | - | 1 | 415 | ||
| AK012743.1 | - | 1 | 415 | |||
| AK134543.1 | - | 1 | 670 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dtna| Gene Symbol: Dtna | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008850.1 | - | 1 | 415 | ||
| AK012743.1 | - | 1 | 415 | |||
| AK134543.1 | - | 1 | 670 | |||