siRNA Details
Gene Symbol: 4930441N18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019607.1 | - | 1 | 356 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714909.1 | - | 1 | 357 | Chr. 4 - 83023799 - 83024245 [+] on Build GRCm38 |
