siRNA Details
| Gene Symbol: 4930441N18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019607.1 | - | 1 | 106 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714909.1 | - | 1 | 106 | Chr. 4 - 83023799 - 83024245 [+] on Build GRCm38 | |

4930441N18Rik
Genomic Map