siRNA Details
| Gene Symbol: C030018G05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021090.1 | - | 1 | 604 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY728185.1 | - | 1 | 602 | Chr. 10 - 108685129 - 108685793 [-] on Build GRCm38 | |

C030018G05Rik
Genomic Map