siRNA Details
| Gene Symbol: C030017B01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046738.1 | - | 1 | 2388 | ||
| BB329116.2 | - | 1 | 46 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BE654522.1 | - | 1 | 230 | Chr. 18 - 40542131 - 40542549 [+] on Build GRCm38 | |

C030017B01Rik
Genomic Map