siRNA Details
Gene Symbol: C030017B01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK046738.1 | - | 1 | 2228 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE654522.1 | - | 1 | 71 | Chr. 18 - 40542131 - 40542549 [+] on Build GRCm38 |
