siRNA Details
| Gene Symbol: Gm19757 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK145340.1 | - | 1 | 236 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY735554.1 | - | 1 | 233 | Chr. 6 - 105483506 - 105492665 [-] on Build GRCm38 | |

Gm19757
Genomic Map